Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0216

Sigma-Aldrich

PF-04725379

≥98% (HPLC)

Synonym(s):

N-((3S)-1-(2-Fluoro-4-(5-((1-methylethylsulfonamido)methyl)-4,5-dihydroisoxazol-3-yl)phenyl)pyrrolidin-3-yl)acetamide, N-((S)-1-(3,5-[3H]-2-Fluoro-4-((S)-5-((1-methylethylsulfonamido)methyl)-4,5-dihydroisoxazol-3-yl)phenyl)pyrrolidin-3-yl)acetamide, PF 04725379

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H27FN4O4S
Molecular Weight:
426.51
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

PF-04725379 is a potent and selective α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) positive allosteric potentiator.
PF-04725379 is a potent and selective AMPAR positive allosteric potentiator.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service