Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0130

Sigma-Aldrich

CP-64434 hydrate

≥98% (HPLC)

Synonym(s):

N′-Desmethylazithromycin hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$115.00
25 MG
$457.00

$115.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$115.00
25 MG
$457.00

About This Item

Empirical Formula (Hill Notation):
C37H70N2O12 · xH2O
CAS Number:
Molecular Weight:
734.96 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$115.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

O.CC[C@H]1OC(=O)[C@H](C)[C@@H](OC2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](OC3O[C@H](C)C[C@H](NC)[C@H]3O)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H70N2O12.H2O/c1-14-26-37(10,45)30(41)23(6)39(12)18-19(2)16-35(8,44)32(51-34-28(40)25(38-11)15-20(3)47-34)21(4)29(22(5)33(43)49-26)50-27-17-36(9,46-13)31(42)24(7)48-27;/h19-32,34,38,40-42,44-45H,14-18H2,1-13H3;1H2/t19-,20-,21+,22-,23-,24+,25+,26-,27+,28-,29+,30-,31+,32-,34+,35-,36-,37-;/m1./s1

InChI key

SREHTTJVVFKIEQ-SXEMRNGDSA-N

Biochem/physiol Actions

CP-064434 is an antibiotic; anti-proliferative HDAC inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service