Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PZ0127

Sigma-Aldrich

CP-74416 methanesulfonate hydrate

≥98% (HPLC)

Synonym(s):

1-Cyclopropyl-7-(1S,4S)-2,5-diazabicyclo[2.2.1]hept-2-yl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid methanesulfonate hydrate, CP-074416 methanesulfonate hydrate, N-desmethyldanofloxacin methanesulfonate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$73.10

$73.10


Check Cart for Availability


Select a Size

Change View
5 MG
$73.10

About This Item

Empirical Formula (Hill Notation):
C18H18FN3O3 · CH3SO3H · xH2O
CAS Number:
Molecular Weight:
439.46 (anhydrous basis)
UNSPSC Code:
41121801
NACRES:
NA.25

$73.10


Check Cart for Availability

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥22 mg/mL

storage temp.

room temp

SMILES string

Fc1cc2c(cc1N4[C@@H]5C[N+H2][C@H](C4)C5)N(C=C(C2=O)C(=O)[O-])C3CC3

InChI

1S/C18H18FN3O3/c19-14-4-12-15(5-16(14)21-7-9-3-11(21)6-20-9)22(10-1-2-10)8-13(17(12)23)18(24)25/h4-5,8-11,20H,1-3,6-7H2,(H,24,25)/t9-,11-/m0/s1

InChI key

LHZDPJRHQVYKPA-ONGXEEELSA-N

Biochem/physiol Actions

CP-74416 is a Danofloxacin primary metabolite; quinolone antibiotic; inhibitor of the bacterial DNA gyrase
CP-74416 is a primary Danofloxacin metabolite. The compound belongs to a group of quinolone antibiotic. CP-74416 is an inhibitor of the bacterial DNA gyrase.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service