Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PRS4989

Sigma-Aldrich

Anti-KLRA1 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Killer cell lectin-like receptor subfamily A, member 1, Anti-Ly49, Anti-Ly49L

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLRA1(10748)

Immunogen

a 13 amino acid peptide near the carboxy-terminus of human KLRA1.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4989.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service