Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PRS4953

Sigma-Aldrich

Anti-NIPSNAP2 (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GBAS, Anti-Glioblastoma amplified sequence, Anti-Non-neuronal SNAP25-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GBAS(2631)

General description

NIPSNAP2 (nipsnap homolog 2) belongs to the NIPSNAP family. It is also known as GBAS (glioblastoma amplified sequence). It is located in the mitochondrial inner membrane space. It is highly expressed in skeletal muscle and heart. GBAS codes for a protein, that has tyrosine phosphorylation sites and a transmembrane domain. It is co-amplified with EGFR (epidermal growth factor receptor gene). It is located on chromosome 7p11.2.

Immunogen

a 14 amino acid peptide near the center of human NIPSNAP2.

Biochem/physiol Actions

NIPSNAP2 (nipsnap homolog 2) plays a major role in transcriptional regulation via L-type Ca2+ channels.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4953.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GBAS, a novel gene encoding a protein with tyrosine phosphorylation sites and a transmembrane domain, is co-amplified with EGFR
Wang XY, et al.
Genomics, 49(3), 448-451 (1998)
De novo 393 kb microdeletion of 7p11.2 characterized by aCGH in a boy with psychomotor retardation and dysmorphic features
Varvagiannis K, et al.
Meta Gene, 2, 274-282 (2014)
NIP-SNAP-1 and -2 mitochondrial proteins are maintained by heat shock protein 60
Yamamoto S, et al.
Biochemical and Biophysical Research Communications, 483(3), 917-922 (2017)
Identification of NIPSNAP1 as a nocistatin-interacting protein involving pain transmission
Okuda-Ashitaka E, et al.
The Journal of Biological Chemistry, 287(13), 10403-10413 (2012)
Regulation of CREB signaling through L-type Ca2+ channels by Nipsnap-2
Brittain JM, et al.
Channels (Austin, Tex.), 6(2), 94-102 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service