Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRS3859

Sigma-Aldrich

Anti-UNG2 (ab1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cyclin O, Anti-Uracil-DNA glycosylase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 39 kDa

species reactivity

rat, human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNO(10309)

General description

UNG2 (uracil-DNA glycosylase) is a multifunctional glycosylase,that is located in the nucleus. It is also known as CCNO (cyclin O)and is located on chromosome 5q11. It is present in the apical cytoplasm of multiciliated cells.

Immunogen

a 14 amino acid peptide from near the amino-terminus of human UNG2.

Biochem/physiol Actions

CCNO (cyclin O) is responsible for the differentiation of multiciliated cell. It participates in the cell cycle and DNA repair. CCNO mutations cause congenital mucociliary clearance disorder. It is considered as an important enzyme in base excision repair (BER).

Linkage

The action of this antibody can be blocked using blocking peptide SBP3859.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vpr-mediated incorporation of UNG2 into HIV-1 particles is required to modulate the virus mutation rate and for replication in macrophages
Chen R, et al.
The Journal of Biological Chemistry, 279(27), 28419-28425 (2004)
Cell cycle-specific UNG2 phosphorylations regulate protein turnover, activity and association with RPA
Hagen L, et al.
The Embo Journal, 27(1), 51-61 (2008)
Cell cycle gene expression networks discovered using systems biology: Significance in carcinogenesis
Scott RE, et al.
Journal of Cellular Physiology, 230(10), 2533-2542 (2015)
Julia Wallmeier et al.
Nature genetics, 46(6), 646-651 (2014-04-22)
Using a whole-exome sequencing strategy, we identified recessive CCNO (encoding cyclin O) mutations in 16 individuals suffering from chronic destructive lung disease due to insufficient airway clearance. Respiratory epithelial cells showed a marked reduction in the number of multiple motile
Structural basis for the recognition of DNA repair proteins UNG2, XPA, and RAD52 by replication factor RPA
Mer G, et al.
Cell, 103(3), 449-456 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service