Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PRS3851

Sigma-Aldrich

Anti-MD-1 (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Lymphocyte antigen 86

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LY86(9450)

General description

MD-1 is a lymphocyte antigen 86 (LY86). The mature peptide is a secretory molecule, has 143 amino acids with no hydrophobic stretches. It has two N-terminal glycosylation sites. Human Md-1 is homologous to mice and chicken Md-1. Out of seven cysteine residues, six is conserved in all three species. MD-1 protein is expressed on the cell surface and lacks transmembrane region. MD-1/Radioprotective 105(RP105) complex is expressed on cell surfaces of B-cells, dendritic cells and macrophages. In human chromosome, the gene MD-1 is localised on 6p25.1.

Immunogen

a 15 amino acid peptide from near the center of human MD-1.

Biochem/physiol Actions

MD-1 associates with Radioprotective 105 (RP105) protein. MD-1 mediates cell surface expression of RP105. Together MD-1 and RP105 are required for lipopolysaccharide (LPS) recognition and signalling. Lack of MD-1 leads to reduced responsiveness to microbial membrane/LPS in B cells. Expression of MD-1 in dendritic cells and macrophages leads to inhibition of TLR4 (Toll like receptor 4)/MD-2 responsiveness to LPS. Methylation of LY86 gene causes obesity and plays a key role in obesity related metabolic disorder.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3851.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Requirement for MD-1 in cell surface expression of RP105/CD180 and B-cell responsiveness to lipopolysaccharide
Nagai Y et al.
Blood, 99(5), 1699-1705 (2002)
DNA methylation of the LY86 gene is associated with obesity, insulin resistance, and inflammation
Su S, et al.
Twin Research and Human Genetics : The Official Journal of the International Society for Twin Studies, 17(3), 183-191 (2014)
Genetics and epigenetics of obesity
Herrera BM, et al.
Maturitas, 69(1), 41-49 (2011)
Y Miura et al.
Blood, 92(8), 2815-2822 (1998-10-09)
RP105 was originally discovered as a mouse B-cell surface molecule that transmits an activation signal. The signal leads to resistance against irradiation-induced apoptosis and massive B-cell proliferation. Recently, we found that mouse RP105 is associated with another molecule, MD-1. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service