Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PRKE

Sigma-Aldrich

Enterokinase Removal Kit

Synonym(s):

PRK-E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160410
NACRES:
NA.56

shipped in

wet ice

storage temp.

2-8°C

General description

Kit is designed to remove all enterokinase (as judged by antigenic or enzymatic activity assays). This is accomplished by binding with immobilized rabbit antibodies to calf intestine enterokinase followed by spin filtration.

Application

For removal of enterokinase by immunoaffinity from fusion protein cleavage reaction mixtures. 100 μl of a 50% slurry of the anti-enterokinase-agarose can remove at least 7 μg of bovine enterokinase or 1 μg of recombinant catalytic subunit.

Preparation Note

Dilute required amount of 20X Wash Buffer 1:20 in distilled water.

Kit Components Only

Product No.
Description

  • 20× Wash buffer 4 mL

  • Disposable spin filters 10 ea

Kit Components Also Available Separately

Product No.
Description
SDS

  • Anti-Enterokinase-agarose 1.5 mL

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai Maekawa et al.
Journal of hypertension, 27(1), 181-189 (2009-01-17)
Prostasin, a glycosylphosphatidylinositol-anchored serine protease, regulates epithelial sodium channel (ENaC) activity. Sodium reabsorption through ENaC in distal nephron segments is a rate-limiting step in transepithelial sodium transport. Recently, proteolytic cleavage of ENaC subunits by prostasin has been shown to activate
Mary Harpen et al.
Journal of virology, 83(21), 10869-10876 (2009-08-28)
As obligatory parasites, viruses co-opt a variety of cellular functions for robust replication. The expression of the nonsegmented negative-strand RNA genome of respiratory syncytial virus (RSV), a significant pediatric pathogen, absolutely requires actin and is stimulated by the actin-regulatory protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service