Key Documents
PQ0101
Supelco
ProteoQwest Colorimetric Western Blotting Kit, TMB Substrate
for Mouse Monoclonal IgG Antibodies
About This Item
UNSPSC Code:
12352200
antibody product type
secondary antibodies
storage temp.
2-8°C
General description
All of the components of this ProteoQwest kit have been extensively tested and optimized.
ProteoQwest Colorimetric Western Blotting Kit, TMB Substrate, is designed for high sensitivity colorimetric detection of as little as 0.15 ng of protein. The colorimetric reaction occurs directly on the PVDF or nitrocellulose membrane; no darkroom or film is needed.
This kit is designed for 25 mini-gel sized (10 × 10 cm) blots. It is possible to use this kit for as many as 45 blots if half the suggested amount of reagents is used.
Legal Information
ProteoQwest is a trademark of Sigma-Aldrich Co. LLC
Not finding the right product?  
Try our Product Selector Tool.
Kit Components Only
Product No.
Description
  • Chemichrome Western Control, (Color Molecular Weight Marker) 200 μL
  • Tris Buffered Saline with 0.05% TWEEN 20, pH 8 25 packets
  • Anti-mouse IgG HRP-conjugated antibody 75 μL
Kit Components Also Available Separately
Product No.
Description
SDS
  • W0138Western Blocker Solution, for HRP detection systems 2 x 400SDS
  • T05653,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for Membranes, ready to use solution 100 mLSDS
Choose from one of the most recent versions:
Certificates of Analysis (COA)