Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PP2363

Sigma-Aldrich

Mammalian Renilla Vector Set

plasmid vectors for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

form

buffered aqueous solution

bacteria selection

ampicillin
kanamycin

origin of replication

pUC (500 copies)

promoter

Promoter name: CMV
Promoter activity: constitutive
Promoter type: mammalian

reporter gene location

2nd promoter
MCS

reporter gene

Renilla luciferase

shipped in

ambient

storage temp.

−20°C

General description

Molecular cloning often benefits from optimizing the vector used for expression.

This pack allows you to determine the best configuration for Renilla luciferase reporter gene and to optimise expression for your specific needs. Each component plasmid contains Renilla luciferase regulated in a different way - in the MCS under the CMV promoter, regulated by your own chosen promoter or under control of an IRES, and three with luciferase regulated by independent promoters either immediately downstream of the MCS (sharing a polyA with the gene inserted into the MCS) or in a different part of the plasmid. Comparing these different strategies for luciferase expression should enable you to identify which is optimum for your particular needs.This plasmid set has been designed to be compatible with a range of cloning techniques. The multiple cloning site contains a range of standard commonly used restriction sites for cloning. Using these sites genes can be inserted using standard cloning methods with DNA ligase. Other methods such as ligase independent cloning (LIC) Gibson Assembly InFusionHD or Seamless GeneArt can also be used and because all of our plasmids are based on the same backbone the same method can be used for cloning into all of our catalogue vectors.

Multiple cloning site notes: There are a few important sites within the MCS. These include the NcoI site the XbaI site and the BsgI and BseRI sites. The NcoI site contains a start codon that is immediately downstream of both a Kozak and Shine-Dalgarno ribosomal binding site. These allow for optimal positioning of genes when the start codon is placed in this location. If this is not required and you wish to use a downstream site for gene cloning you can remove the NcoI site by cleaving the plasmid with KpnI. The XbaI site contains a stop codon. This stop codon is positioned in a specific position in relation to the BsgI and BseRI sites that are immediately downstream. When either BseRI or BsgI cleave the plasmid they produce a TA overhang from the stop codon in the XbaI site that is compatible with all of our peptide tag plasmids cut with the same sites. BseRI and BsgI sites are non-palindromic and cleave a defined number of bases away from their binding site. Whenever we clone a gene into our multiple cloning site we always position the start and stop codon in the same positions in the MCS. If the start and ends of the genes are not compatible with NcoI and XbaI we extend the sequence to the nearest external sites but keep the start and stop codons locations consistent.

Transcription Termination:<p>These plasmids contains three alternative transcription terminators for mammalian bacterial and bacteriophage (T7) expression. This means that only the promoter needs to be changed to alter the expression system you are using. We sell multiple promoters that can be used in each of these systems. The presence of each terminator does not reduce expression in the alternative systems.

Intellectual Property Status: According to our IP-friendly policy this plasmid is sold free of reach-through rights and can be used to make commercial products. However the plasmid itself (or derivatives) cannot be sold.

Sequence

To view sequence information for this product, please visit the product page

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Legal Information

These plasmids are sold free of reach-through rights and can be used to make commercial products. However the plasmids themselves (or derivatives) cannot be sold.

Kit Components Also Available Separately

Product No.
Description
SDS

  • OGS103PSF-CMV-RLUC - CMV RENILLA LUCIFERASE PLASMID, plasmid vector for molecular cloningSDS

  • OGS16PSF-CMV-RSV-RLUC ASCI - RENILLA LUCIFERASE PLASMID, plasmid vector for molecular cloningSDS

  • OGS286PSF-CMV-FMDV-RLUC - FMDV IRES RENILLA LUCIFERASE VECTOR, plasmid vector for molecular cloningSDS

  • OGS237PSF-PA-PROMMCS-RLUC - NO PROMOTER (MCS) RENILLA PLASMID, plasmid vector for molecular cloningSDS

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service