Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLX50

Sigma-Aldrich

GenElute Plasmid Maxiprep Kit

sufficient for 50 purifications

Synonym(s):

Gen Elute, GenElute Plasmid Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 50 purifications

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

The GenElute Plasmid Maxiprep Kit offers a simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cultures. By combining silica-binding technology and the convenience of a spin column format, up to 1.2 mg of plasmid DNA can be recovered from 25-200 ml of Luria Broth (LB) in about 45 minutes. Note that actual yield and optimal volume of culture to use depend on the plasmid and the culture medium.

An overnight recombinant E. coli culture is harvested by centrifugation and subjected to a modified alkaline-SDS lysis procedure followed by adsorption of the DNA onto silica in the presence of high salts. Contaminants are then removed by a spin-wash step. Finally, the bound DNA is eluted in water or Tris-EDTA buffer.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characterisation of mutations in GroES that allow GroEL to function as a single ring.
Liu H
Febs Letters, 583(14), 2365-2371 (2009)
Martina Herrmann et al.
Applied and environmental microbiology, 75(10), 3127-3136 (2009-03-24)
Communities of ammonia-oxidizing archaea (AOA) and bacteria (AOB) in freshwater sediments and those in association with the root system of the macrophyte species Littorella uniflora, Juncus bulbosus, and Myriophyllum alterniflorum were compared for seven oligotrophic to mesotrophic softwater lakes and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service