Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLAK0787

Sigma-Aldrich

53BP1 and BRCA1 PLA® Antibody Kit

Powered by Bethyl Laboratories, Inc.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

technique(s)

proximity ligation assay: suitable (indirect)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Application

Product PLAK0787, Powered by Bethyl Laboratories, has been pre-qualified for use in proximity ligation assay (PLA®). PLA® technology enables visualization of protein interactions in fixed cells and tissue samples while under endogenous protein expression levels.
The secondary-conjugate (indirect) method of PLA® requires the use of a pair of Duolink® probes, one PLUS and one MINUS, and your choice of Detection Reagents. Accessories include Wash Buffers and Mounting Media.

Linkage

This kit contains antibody PLA0303 and antibody PLA0185, which have been pre-qualified as an antibody pair for use in Proximity Ligation Assay. To view all Duolink products, visit sigma-aldrich.com/duolink.

Analysis Note

The antibody for 53BP1 (PLA0303) is used at dilution 1:10,000 and the antibody for BRCA1 (PLA0185) is used at dilution 1:10,000 for PLA applications.

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service