PLAK0462
HIF1-alpha and p300 PLA® Antibody Kit
Powered by Bethyl Laboratories, Inc.
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
technique(s)
proximity ligation assay: suitable (indirect)
shipped in
wet ice
Storage temp.
2-8°C
target post-translational modification
unmodified
Application
Product PLAK0462, Powered by Bethyl Laboratories, has been pre-qualified for use in proximity ligation assay (PLA®). PLA® technology enables visualization of protein interactions in fixed cells and tissue samples while under endogenous protein expression levels.
The secondary-conjugate (indirect) method of PLA® requires the use of a pair of Duolink® probes, one PLUS and one MINUS, and your choice of Detection Reagents. Accessories include Wash Buffers and Mounting Media.
Linkage
This kit contains antibody PLA0304 and antibody PLA0096, which have been pre-qualified as an antibody pair for use in Proximity Ligation Assay. To view all Duolink products, visit sigma-aldrich.com/duolink.
Analysis Note
The antibody for HIF1-alpha (PLA0304) is used at dilution 1:10,000 and the antibody for p300 (PLA0096) is used at dilution 1:1,000 for PLA applications.
Legal Information
Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service