Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0298

Sigma-Aldrich

Goat anti-RPA32 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

32kDa, REPA2, RF-A protein 2, RP-A p32, RP-A p34, RPA32, replication factor A protein 2, replication protein A 34 kDa subunit, replication protein A2, replication protein A2 (32kD)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$477.00

$477.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$477.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$477.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:500-1:2,000
western blot: 1:500- 1:2,500

accession no.

NP_002937.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... RPA32(6118)

Immunogen

The epitope recognized by PLA0298 maps to a region between residue 220 and 270 of human Replication Protein A2 (32kD) using the numbering given in entry NP_002937.1 (GeneID 6118).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Replication protein A (RPA) is a multisubunit complex that carries out DNA mismatch repair (MMR) in association with MSH2 (a subunit of human MutS heterodimers), MLH1 (a subunit of human MutL heterodimers), PCNA, and DNA polymerase-delta. RPA is composed of a heterotrimer that includes subunits of 70 kDa (RPA1), 32kDa (RPA2), and 14kDa (RPA3). RPA2, the 32kDa subunit, is phosphorylated by the cdc2 family of kinases when cells enter S-phase and in response to DNA damage by ATM, ATR, and DNA-PK.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service