Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0285

Sigma-Aldrich

Rabbit anti-Phospho MEK1(T286) Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CFC3, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK 1, MAPKK1, MEK 1, MEK1, MKK1, PRKMK1, kinase 1 (MAP kinase kinase 1), mitogen-activated, protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:1,000-1:5,000
immunoprecipitation (IP): 2- 10 μg/mg

accession no.

NP_002746.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... MAP2K1(5604)

Immunogen

The epitope recognized by PLA0285 maps to a region of human MAPK/ERK kinase 1 (dual specificity mitogen-activiated protein kinase kinase 1) surrounding threonine 286 using the numbering given in entry NP_002746.1 (Gene ID 5604) when the threonine residue is phosphorylated.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

MEK1 is an activator and kinase for MAP and ERK kinases. It is a serine/threonine kinase essential to the MAP kinase signal transduction pathway which is involved in a multitude of cellular processes such as proliferation, differentiation, gene activation, and development. MEK1 displays 85% identity with MEK2. Although MEK1 and MEK2 have been found to be functionally similar, they may serve non-redundant functions. Both are required for cellular proliferation and may play a role in tumorigenesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service