Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0284

Sigma-Aldrich

Rabbit anti-ELK1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ELK1, ETS-like gene 1, member of ETS oncogene family, tyrosine kinase (ELK1) oncogene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_005220.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... ELK1(2002)

Immunogen

The epitope recognized by PLA0284 maps to a region between residue 378 and 428 of human ETS-like Gene 1 using the numbering given in entry NP_005220.2 (GeneID 2002).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

ELK1 is a member of the Ets family of transcription factors and of the ternary complex factor (TCF) subfamily. Proteins of the TCF subfamily form a ternary complex by binding to the the serum response factor and the serum reponse element in the promoter of the c-fos proto-oncogene. ELK1 is a nuclear target for the ras-raf-MAPK signaling cascade [taken from NCBI Entrez Gene (Gene ID: 2002)].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service