Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0268

Sigma-Aldrich

Rabbit anti-YY1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

DELTA, INO80 complex subunit S, INO80S, NF-E1, UCRBP, YIN-YANG-1, YY-1, Yin and Yang 1 protein, Yin and yang 1, delta transcription factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_003394.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... YY1(7528)

Immunogen

The epitope recognized by PLA0268 maps to a region between residue 50 and 100 of human Yin and Yang 1 Protein using the numbering given in entry NP_003394.1 (GeneID 7528).

Physical form

Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide

Other Notes

YY1 (ying and yang 1) is a multifunctional transcription factor that exhibits positive and negative control on a large number of cellular and viral genes by binding to sites overlapping the transcription start site. YY1 may play an important role in development and differentiation. The function of YY1 as an activator or a repressor is specified by the presence of other proteins. For example it acts as a repressor in absence of adenovirus E1A protein but as an activator in its presence.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service