Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0258

Sigma-Aldrich

Rabbit anti-CDC7 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CDC7 (cell division cycle 7; S. cerevisiae; homolog)-like 1, CDC7 cell division cycle 7 (S. cerevisiae), CDC7-related kinase, CDC7L1, HsCdc7, Hsk1, cell division cycle 7 (S. cerevisiae), cell division cycle 7 homolog, cell division cycle 7 homolog (S. cerevisiae), cell division cycle 7-like protein 1, huCdc7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_003494.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... CDC7(8317)

Immunogen

The epitope recognized by PLA0258 maps to a region between residue 524 and 574 of human cell division cycle 7 using the numbering given in entry NP_003494.1 (GeneID 8317).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Cell division cycle 7-related protein kinase (CDC7) is critical for the activation and firing of replication origins during S-phase. CDC7 kinase activity is activated and regulated by Dbf4. Substrates of CDC7 include the MCM (mini-chromosome maintenance) proteins, DNA polymerase alpha, Cdc45, Orc4, geminin and SV40 T-antigen. CDC7 has also been implicated to play a role in the DNA damage response.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service