Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0235

Sigma-Aldrich

Rabbit anti-ERK1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

ERK-1, ERK1, ERT2, HS44KDAP, HUMKER1A, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, PRKM3, extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, insulin-stimulated MAP2 kinase, microtubule-associated protein 2 kinase, p44-ERK1, p44-MAPK, p44erk1, p44mapk

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$504.00

$504.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$504.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$504.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:100-1:500
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_002737.2

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... ERK1(5595)

Immunogen

The epitope recognized by PLA0235 maps to a region between residue 329 and 379 of human extracellular signal-related kinase 1 (mitogen-activated protein kinase 3) using the numbering given in entry NP_002737.2 (GeneID 5595).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Extracellular signal-regulated kinase 1 (ERK1/p44) is also known as mitogen-activated protein kinase 3 (MAPK3). ERK1 is part of a signal transduction pathway that begins with the small GTPase Ras and continues through a cascade of protein kinases that include Raf and MEK. Activation of this signaling pathway influences many cellular processes such as proliferation, differentiation and survival. ERK1 is very similar in sequence to ERK2/p42 and the activities of both ERK1 and ERK2 are usually considered together as total ERK activity. More recent evidence has shown that ERK1 and ERK2 are not functionally redundant and may have very different and specific roles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emmanuel Prikas et al.
The EMBO journal, 41(18), e10242-e10242 (2022-08-23)
Microtubule-associated protein tau is a central factor in Alzheimer's disease and other tauopathies. However, the physiological functions of tau are unclear. Here, we used proximity-labelling proteomics to chart tau interactomes in primary neurons and mouse brains in vivo. Tau interactors map

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service