Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0225

Sigma-Aldrich

Rabbit anti-IQGAP1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

HUMORFA01, KIAA0051, RasGAP-like with IQ motifs, SAR1, p195

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:100-1:500
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_003861.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... IQGAP1(8826)

Immunogen

The epitope recognized by PLA0225 maps to a region between residue 1 and 50 of human motif containing GTPase activating protein 1 using the numbering given in entry NP_003861.1 (GeneID 8826).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

IQGAP1 (IQ motif containing GTPase activating protein 1) interacts with the Rho-family GTPases, CDC42 and RAC1. It has been shown to localize to sites of cell-cell contact and regulate cadherin-mediated cell-cell adhesion. In addition to its role in cell adhesion, it is also involved in cell polarization and migration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service