Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0220

Sigma-Aldrich

Rabbit anti-CTF18 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

C16orf41, C321D2.2, C321D2.3, C321D2.3 (novel protein), C321D2.4, C321D2.4 (novel protein), CHL12, CTF18, Ctf18, RUVBL, chromosome 16 open reading frame 41, chromosome transmission fidelity factor 18 homolog, chromosome transmission fidelity factor 18 homolog (S. cerevisiae), hCTF18, homolog of yeast CHL12, some homology with holliday junction DNA helicase RUVB like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:100-1:500
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:1,000- 1:5,000

accession no.

NP_071375.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... CTF18(63922)

Immunogen

The epitope recognized by PLA0220 maps to a region between residue 25 and 75 of human chromosome transmission fidelity factor 18 using the numbering given in entry NP_071375.1 (GeneID 63922).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

CTF18 (Chromosome Transmission Fidelity protein 18 homolog) is part of the CTF18-RFC (Replication Factor C) complex composed of CTF18, CTF8, DCC1, and four RFC subunits. This complex has been shown to be important for the establishment of sister chromatid cohesion and functions as a proliferating cell nuclear antigen (PCNA) loader for DNA synthesis.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service