PLA0203
Rabbit anti-cAbl Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
ABL, Abelson murine leukemia viral oncogene homolog 1, Abelson tyrosine-protein kinase 1, JTK7, bcr/abl, bcr/c-abl oncogene protein, c-ABL, c-ABL1, c-abl oncogene 1, non-receptor tyrosine kinase, p150, proto-oncogene c-Abl, proto-oncogene tyrosine-protein kinase ABL1, receptor tyrosine kinase, v-abl, v-abl Abelson murine leukemia viral oncogene homolog 1
About This Item
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
species reactivity
human
technique(s)
immunofluorescence: 1:100-1:1,000
immunohistochemistry: 1:100- 1:1,000
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:2,000- 1:10,000
accession no.
NP_005148.2
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... cAbl(25)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service