Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0189

Sigma-Aldrich

Rabbit anti-CCDC131 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

C3H1-type containing, CCDC131, CSRC2, KIAA0546, MGC23401, PSRC2, coiled-coil domain containing 131, coiled-coil domain-containing protein 131, proline/serine-rich coiled-coil 2, proline/serine-rich coiled-coil protein 2, zinc finger

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$504.00

$504.00


Check Cart for Availability


Select a Size

Change View
100 μL
$504.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$504.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_659419.3

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... CCDC131(196441)

Immunogen

The epitope recognized by PLA0189 maps to a region between residue 300 and 350 of human coiled-coil domain containing 131 using the numbering given in entry NP_659419.3 (GeneID 196441).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

CCDC131 (coiled-coil domain containing protein 131) is an uncharacterized proline/serine rich coiled-coil protein that bears 6 HAT repeats- and 5 TPR repeats. Coiled-coil proteins exhibit tertiary structures made up of a number of highly organized alpha helices wound around each other to mediate oligomerization. HAT (half a TPR) repeats form amphipathic alpha-helices and are similar to TPR repeats which are 34 amino acid repeat motifs which form alpha helical structures.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service