Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0187

Sigma-Aldrich

Rabbit anti-BAF53A Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

53 kDa BRG1-associated factor A, ARPN-BETA, Actl6, Arp4, BAF complex 53 kDa subunit, BAF53, BAF53A, BRG1-associated factor, BRG1-associated factor 53A, INO80 complex subunit K, INO80K, actin-like 6A, actin-related protein 4, actin-related protein Baf53a, arpNbeta, hArpN beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:100-1:500
western blot: 1:2,000- 1:10,000

accession no.

NP_004292.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... BAF53A(86)

Immunogen

The epitope recognized by PLA0187 maps to a region between residue 25 and 75 of human BRG1-associated factor, 53 kDa subunit A using the numbering given in entry NP_004292.1 (GeneID 86).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

BAF53A is a 53kd subunit of the BAF (BRG1/brm-associated factor) complex, a SWI/SNF chromatin remodeling and histone acetyltransferase complex that functions in the regulation of gene transcription. In the BAF complex, BAF53A is important to the ATPase activity of SMARCA4/BRG1 and the association of the BAF complex with chromatin. BAF53A also appears to be important to the formation of higher-order chromatin structure and has been shown to associate with mitotic chromosomes. Recently, BAF53A has been found to interact with p53 and repress p53-mediated gene transcription in a BRG1-independent manner.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service