Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0180

Sigma-Aldrich

Rabbit anti-GTF3C1/TFIIIC220 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

220kD ), TF3C-alpha, TFIIIC, TFIIIC 220 kDa subunit, TFIIIC box B-binding subunit, TFIIIC220, TFIIICalpha, alpha 220kDa, general transcription factor IIIC, polypeptide 1, polypeptide 1 (alpha subunit, polypeptide 1 (alpha subunit), transcription factor IIIC 220 kDa subunit, transcription factor IIIC subunit alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:2,000
western blot: 1:2,000- 1:10,000

accession no.

NP_001511.2

shipped in

wet ice

storage temp.

2-8°C

Gene Information

Immunogen

The epitope recognized by PLA0180 maps to a region between residue 2059 and 2109 of human general transcription factor IIIC, polypeptide 1 (general transcription factor IIIC, 220kDa subunit) using the numbering given in entry NP_001511.2 (GeneID 2975).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

General transcription factor 3C polypeptide 1 (GTF3C1/TFIIIC220) is a subunit of the TFIIIC DNA-binding factor required for transcription of tRNA and 5S rRNA genes by RNA polymerase III. Human TFIIIC is composed of six subunits: TFIIIC220, TFIIIC110, TFIIIC102, TFIIIC90 and TFIIIC63, and TFIIIC35. The functional contribution of GTF3C1/TFIIIC220 to the TFIIIC complex has not been fully determined.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin Van Bortle et al.
Nature communications, 13(1), 3007-3007 (2022-06-01)
RNA polymerase III (Pol III) includes two alternate isoforms, defined by mutually exclusive incorporation of subunit POLR3G (RPC7α) or POLR3GL (RPC7β), in mammals. The contributions of POLR3G and POLR3GL to transcription potential has remained poorly defined. Here, we discover that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service