Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0171

Sigma-Aldrich

Rabbit anti-SMRT Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CTG repeat protein 26, CTG26, N-CoR2, SMAP270, SMRT, SMRTE, SMRTE-tau, Silencing mediator of retinoic acid and thyroid hormone receptor, T3 receptor-associating factor, TNRC14, TRAC, TRAC-1, TRAC1, nuclear receptor co-repressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, thyroid-

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$477.00

$477.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$477.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$477.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

AAD20946.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... SMRT(9612)

Immunogen

The epitope recognized by PLA0171 maps to a region between residue 1525 and 1575 of human silencing mediator of retinoic acid and thyroid hormone receptor alpha using the numbering given in entry AAD20946.1 (GeneID 9612).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

SMRT (silencing mediator of retinoid and thyroid hormone receptor) mediates gene repression by interacting with nuclear receptors such as the thyroid hormone receptor, retinoic acid receptor, progesterone receptor, androgen receptor, estrogen receptor, and glucocorticoids receptor. SMRT mediates repression by associating with SIN3A/B and the histone deacetylases, HDAC1 and HDAC2, and promoting the formation of repressive chromatin structures.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiumei Zhang et al.
PloS one, 14(5), e0217181-e0217181 (2019-05-24)
MiR-873/CDK3 has been shown to play a critical role in ERα signaling and tamoxifen resistance. Thus, targeting this pathway may be a potential therapeutic approach for the treatment of ER positive breast cancer especially tamoxifen resistant subtype. Here we report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service