Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0157

Sigma-Aldrich

Rabbit anti-Sin1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

JC310, MEKK2-interacting protein 1, MGC2745, MIP1, SAPK-interacting protein 1, SIN1, SIN1b, SIN1g, TORC2 subunit MAPKAP1, mSIN1, mitogen-activated protein kinase 2-associated protein 1, ras inhibitor MGC2745, stress-activated map kinase interacting protein 1, stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$477.00

$477.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$477.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$477.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunohistochemistry: 1:1,000- 1:5,000
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_001006618.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Sin1(79109)

Immunogen

The epitope recognized by PLA0157 maps to a region between residue 470 and the C-terminus (residue 522) of human stress-activated map kinase interacting protein 1 using the numbering given in entry NP_001006618.1 (GeneID 79109).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Stress-activated map kinase-interacting protein 1 (Sin1) is highly similar to the yeast stress- activated protein kinase SIN1. Sin1 is an essential mTORC2 subunit that is required to act as the PDK2 that phosphorylates Akt/PKB at Ser 473. Sin1 has been implicated as a scaffolding protein. Sin1 is required for rictor binding in the mTORC2 complex. Sin1 has also been shown to be important to the SAPK (stress-activated kinase) signaling pathway and in the nucleus may act as a scaffold between ATF-2 and p38 to facilitate p38-induced phosphorylation of ATF-2. Sin1 is also found to form a complex with c-Jun N-terminal kinase (JNK) and may similarly function as a scaffolding protein in JNK signaling.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service