PLA0133
Rabbit anti-PTEN Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
10q23del, 4, 5-trisphosphate 3-phosphatase and dual-specificity protein phosphatase PTEN, BZS, CWS1, DEC, GLM2, MHAM, MMAC1, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, PTEN1, Phosphatase and tensin homolog, TEP1, mitochondrial PTENalpha, mitochondrial phosphatase and tensin protein alpha, mutated in multiple advanced cancers 1, phosphatase and tensin-like protein, phosphatidylinositol-3
About This Item
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
species reactivity
human, mouse
technique(s)
immunohistochemistry: 1:100- 1:500
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000- 1:10,000
accession no.
NP_000305.2
UniProt accession no.
shipped in
wet ice
Storage temp.
2-8°C
Gene Information
rabbit ... PTEN(5728)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service