Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0104

Sigma-Aldrich

Rabbit anti-EWS Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

EWS, EWS RNA-binding protein 1, EWS RNA-binding protein variant 6, EWS oncogene, EWS-FLI1, Ewing sarcoma breakpoint region 1, Ewing sarcoma breakpoint region 1 protein, Ewings sarcoma EWS-Fli1 (type 1) oncogene, bK984G1.4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:2,000-1:10,000
immunoprecipitation (IP): 1-5 μg/mg
western blot: 1:10,000- 1:20,000

accession no.

Q01844

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... EWS(2130)

Immunogen

The epitope recognized by PLA0104 maps to a region between residues 100 and 150 of human Ewing sarcoma breakpoint region 1 using the numbering given in SwissProt entry Q01844 (GeneID 2130).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

EWS is a member of a family of RNA-binding proteins that may serve as adapters that bridge RNA polymerase II complex and RNA splicing factors. Translocations and fusions between EWS and the ETS transcription factors promote oncogenesis and are associated with Ewing′s family tumors.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service