Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0103

Sigma-Aldrich

Rabbit anti-CDK7 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

39 KDa protein kinase, CAK, CAK1, CDK-activating kinase 1, CDKN7, HCAK, MO15, STK1, Serine/threonine-protein kinase 1, TFIIH basal transcription factor complex kinase subunit, Xenopus laevis, cdk-activating kinase), cell division protein kinase 7, cyclin-dependent kinase 7 (MO15 homolog, cyclin-dependent kinase 7 (homolog of Xenopus MO15 cdk-activating kinase), homolog of Xenopus MO15 Cdk-activating kinase, kinase subunit of CAK, p39 Mo15, p39MO15, serine/threonine kinase stk1, serine/threonine protein kinase 1, serine/threonine protein kinase MO15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: 1:100-1:500
immunohistochemistry: 1:500-1:2,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000

accession no.

P50613

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... CDK7(1022)

Immunogen

The epitope recognized by PLA0103 maps to a region between residues 300 and the C-terminus (residue 346) of human Cyclin-Dependent Kinase 7 using the numbering given in Swiss-Prot entry P50613 (GeneID 1022).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Cyclin-dependent kinase 7 (CDK7) is a member of the CDK family of protein kinases that regulate cell cycle progression. CDK7 is the catalytic subunit in a trimeric complex with cyclin H and MAT1 that functions as a Cdk-activating kinase (CAK). CDK7 activates the kinase activities of CDK1, CDK2, CDK4, and CDK6 by threonine phosphorylation. In addition to its cell cycle function CDK7 has also been found to be an essential component of the transcription factor TFIIH. Several substrates of CDK7 are involved in transcription, further indicating a role in transcription.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service