Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

PLA0094

Sigma-Aldrich

Rabbit anti-SRC2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

GRIP1, KAT13C, NCoA-2, SRC2, TIF2, bHLHe75, class E basic helix-loop-helix protein 75, glucocorticoid receptor-interacting protein-1, hTIF2, p160 steroid receptor coactivator 2, transcriptional intermediary factor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 1-4 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_006531.1

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... SRC2(10499)

Immunogen

The epitope recognized by PLA0094 maps to a region between residue 1400 the C-terminus (residue 1464) of human Nuclear Receptor Coactivator 2 using the numbering given in entry NP_006531.1 (GeneID 10499).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Steroid receptor coactivator 2 (SRC2) also known as TIF2 belongs to the SRC family of proteins that interacts with nuclear receptors to facilitate transcription. The 3 homologous members of this family are SRC-1 (NCoA-1), SRC-2 (GRIP1, TIF2, or NcoA-2), and SRC-3 (p/CIP, RAC3, ACTR, AIB1, or TRAM-1). SRC2 has been found to serve a coactivator for PPAR-gamma and to play an important role in lipid metabolism.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service