Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0092

Sigma-Aldrich

Rabbit anti-MAD1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

HsMAD1, MAD1, MAD1 (mitotic arrest deficient; yeast; homolog)-like 1, MAD1 mitotic arrest deficient-like 1, MAD1-like protein 1, PIG9, TP53I9, TXBP181, hMAD1, homolog-like 1, mitotic arrest deficient 1-like protein 1, mitotic checkpoint MAD1 protein homolog, mitotic-arrest deficient 1, tax-binding protein 181, tumor protein p53 inducible protein 9, yeast

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$504.00

$504.00


Check Cart for Availability


Select a Size

Change View
100 μL
$504.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$504.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 1-4 μg/mg
western blot: 1:5,000- 1:15,000

accession no.

Q9Y6D9

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... MAD1(8379)

Immunogen

The epitope recognized by PLA0092 maps to a region between residues 1 and 50 of human Mitotic arrest deficient-like 1 using the numbering given in Swiss-Prot entry Q9Y6D9 (GeneID 8379).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

MAD1 (Mitotic Arrest Deficiency 1) is a component of the mitotic spindle-assembly checkpoint that ensures that all chromosomes are aligned at the metaphase plate before the onset of anaphase. MAD1 functions as a homodimer that interacts with and regulates MAD2. MAD1 may play a role in tumor suppression.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirohisa Kyogoku et al.
Developmental cell, 41(3), 287-298 (2017-05-10)
Chromosome segregation during meiosis in oocytes is error prone. The uniquely large cytoplasmic size of oocytes, which provides support for embryogenesis after fertilization, might be a predisposing factor for meiotic errors. However, this hypothesis remains unproven. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service