PLA0084
Rabbit anti-FOXO1a Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
Drosophila, FKH1, FKHR, FOXO1A, Forkhead in rhabdomyosarcoma, forkhead, forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, homolog of, in rhabdomyosarcoma
About This Item
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
species reactivity
human
technique(s)
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000-1:10,000
accession no.
Q12778
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... FOXO1a(2308)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service