Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

PLA0064

Sigma-Aldrich

Rabbit anti-MCM5 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CDC46, CDC46 homolog, MCM5 minichromosome maintenance deficient 5, P1-CDC46, cell division cycle 46, cell division cycle 46 (S. cerevisiae), minichromosome maintenance deficient (S. cerevisiae) 5 (cell division cycle 46), minichromosome maintenance deficient 5 (cell division cycle 46)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:500- 1:2,000
immunoprecipitation (IP): 2- 4 μg/mg
western blot: 1:5,000-1:20,000

accession no.

NP_006730.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... MCM5(4174)

Immunogen

The epitope recognized by PLA0064 maps to a region between residues 1 and 50 of human Minichromosome Maintenance 5 using the numbering given in entry NP_006730.2 (GeneID 4174).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

The MCM (Mini-Chromosome Maintenance) complex is a key component of the pre-replication complex involved in replication licensing which restricts DNA replication to only once per cell cycle. The MCM complex is a heterohexamer of MCM2, MCM3, MCM4, MCM5, MCM6, and MCM7. MCM5 can form a subcomplex with MCM3. MCM5 also appears to play a role in transcription. MCM5 has been shown to interact with the signal transducer and activator of transcription 1 (STAT1) and demonstrated to be essential for STAT1 target gene transcription.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maïté Leturcq et al.
Cellular and molecular life sciences : CMLS, 75(23), 4321-4339 (2018-08-03)
O-GlcNAcylation of proteins is governed by O-GlcNAc transferase (OGT) and O-GlcNAcase (OGA). The homeostasis of O-GlcNAc cycling is regulated during cell cycle progression and is essential for proper cellular division. We previously reported the O-GlcNAcylation of the minichromosome maintenance proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service