Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0058

Sigma-Aldrich

Rabbit anti-Rad50 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

NBSLD, RAD50 (S. cerevisiae) homolog, RAD50 homolog, RAD50 homolog (S. cerevisiae), RAD50-2, RAD502, double strand break repair protein, hRad50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 1- 4 μg/mg
western blot: 1:1,000-1:5,000

accession no.

NP_005723.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... Rad50(10111)

Immunogen

The epitope recognized by PLA0058 maps to a region between residues 50 and 100 of human Rad50 using the numbering given in entry NP_005723.2 (GeneID 10111).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Rad50 is a component of the Mre11-Rad50-Nbs1 (MRN) complex that repairs DNA double strand breaks during DNA damage and homologous recombination and activates the S-phase checkpoint. Rad50 shows structural similarity to the structural maintenance of chromosomes (SMC) family of proteins. Rad50 has been shown to directly interact with BRCA2, TERF2IP, and ATM.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service