Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0027

Sigma-Aldrich

Goat anti-Menin Antibody, Affinity Purified

Synonym(s):

MEAI, SCG2, menin, multiple endocrine neoplasia I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$504.00

$504.00


Check Cart for Availability


Select a Size

Change View
100 μL
$504.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$504.00


Check Cart for Availability

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:1,000- 1:10,000

accession no.

NP_000235.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... Menin(4221)

Immunogen

The epitope recognized by PLA0027 maps to a region between residue 575 and the C-terminus (residue 615) of human Multiple Endocrine Neoplasia 1 using the numbering given in entry NP_000235.2 (GeneID 4221).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

Menin is the gene product of the putative tumor suppressor MEN1 associated with the syndrome termed multiple endocrine neoplasia type 1 that may function in the regulation of DNA synthesis or the response to DNA damage. Menin has been shown to interact with JunD, a member of the AP-1 heterodimer and inhibit transcription. Menin has also been shown to interact with Pem, FANCD2, RPA, NMMHCII-A, GFAP, vimentin, Hsp70, NfkappaB proteins, Smads, mSin3A and is part of the MLL2 and MLL1 Set1-like histone H3 K4 methyltransferase complex where it interacts directly with ASH2, Rbbp5, and WDR5. As part of the MLL complexes, menin may mediate its tumor suppressor activity via the regulation of histone methylation of HOX and CDK inhibitor genes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service