Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PLA0020

Sigma-Aldrich

Rabbit anti-SMC4 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CAP-C, CAPC, SMC protein 4, SMC-4, SMC4 (structural maintenance of chromosomes 4; yeast)-like 1, SMC4 structural maintenance of chromosomes 4-like 1, SMC4 structural maintenance of chromosomes 4-like 1 (yeast), SMC4L1, XCAP-C homolog, chromosome-associated polypeptide C, hCAP-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunocytochemistry: 1:200-1:400
immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:1,000- 1:10,000

accession no.

NP_005487.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... SMC4(10051)

Immunogen

The epitope recognized by PLA0020 maps to a region between residue 1225 and the C-terminus (residue 1288) of human Structural Maintenance of Chromosomes 4 using the numbering given in entry NP_005487.2 (GeneID 10051).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

SMC (Structural maintenance of chromosomes) proteins are part of the cohesin and condensin multisubunit complexes that play a critical role in chromosome organization, segregation, gene regulation, and DNA repair. The core condensin complex includes a heterodimer of SMC2 and SMC4 that functions to create positively supercoiled loops for chromatin condensation and metaphase chromatid shaping. A function for condensin in the DNA damage checkpoint response and gene repression has also been suggested.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service