Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PG500200

Supelco

Controlled pore glass–uncoated

nominal diameter 500 Å, 120-200 mesh

Synonym(s):

Glass, controlled-pore

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41115716

form

powder

technique(s)

LPLC: suitable

matrix

Silica

matrix active group

polymer

particle size

120-200 mesh

pore size

500 Å nominal diameter

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service