Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PG1400120

Supelco

Controlled pore glass–uncoated

nominal diameter 1,400 Å, 80-120 mesh

Synonym(s):

Glass, controlled-pore

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41115716

form

powder

technique(s)

LPLC: suitable

matrix

Silica

matrix active group

polymer

particle size

80-120 mesh

pore size

1,400 Å nominal diameter

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service