PG120120
Controlled pore glass–uncoated
nominal diameter 120 Å, 80-120 mesh
Synonym(s):
Glass, controlled-pore
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
41115716
form
powder
technique(s)
LPLC: suitable
matrix
Silica
matrix active group
polymer
particle size
80-120 mesh
pore size
120 Å nominal diameter
separation technique
size exclusion (SEC)
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service