Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9993

Sigma-Aldrich

Anti-PKR (KT-16) antibody produced in rabbit

1-2 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Interferon-induced, double-stranded RNA-activated protein kinase (KT-16)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 68 kDa

species reactivity

rat, human

concentration

1-2 mg/mL

technique(s)

western blot: 1:2,000 using whole cell extract of the human epitheloid carcinoma HeLa cell line induced with interferon-γ.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide near the C-terminus of human PKR (amino acids 522-537).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service