Key Documents
P9934
Pipette tips, 200 μL
With aerosol barrier, usable capacity 30μL, sterile
About This Item
UNSPSC Code:
41121601
NACRES:
NB.01
material
colorless
sterility
sterile
packaging
pack of 96 ea (Trays)
case of 960 ea (10 trays of 96 tips)
maximum volume
200 μL
General description
Inert, bonded fiber barrier coalesces airborne contaminants so they do not enter pipette barrel.
Placement of barrier reduces usable capacity to 30 μl.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)