Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9868

Sigma-Aldrich

Anti-Protein Phosphatase 5 (PP5) antibody produced in rabbit

buffered aqueous solution, fractionated antiserum, pkg of ~100 μg, 1 mg/mL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58 kDa

species reactivity

human

packaging

pkg of ~100 μg

concentration

1 mg/mL

technique(s)

western blot: 5-10 μg/mL using A549 or MCF7 cell lysates

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PPP5C(5536)

General description

Ser/Thr protein phosphatase type 5 (PP5) is a 58kD protein present in human and is localized in nucleus which plays an essential role in mitosis and in the regulation of RNA biogenesis. It also has a pivotal role in cell proliferation and facilitates neoplastic transformation. Rabbit Anti-Protein Phosphatase 5 (PP5) antibody can identify human PP5 and reacts specifically with protein serine threonine phosphatase 5 (58 kD).

Immunogen

synthetic peptide derived from the human phosphatase 5 protein.

Application

Anti-Protein Phosphatase 5 (PP5) antibody can be used in western blotting using A549 or MCF7 cell lysates.

Physical form

Solution in phosphate buffered saline containing 0.08% sodium azide. The amount of the reagent is sufficient for 10 blots.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Chinkers
Trends in endocrinology and metabolism: TEM, 12(1), 28-32 (2001-01-04)
Protein phosphatase 5 (PP5) possesses unique biochemical properties, which include its tetratricopeptide repeat (TPR) targeting/regulatory domain and its ability to be activated by lipids. PP5 has been studied as a paradigm for TPR protein structure and function. Roles for PP5
Z Zuo et al.
Biochemistry, 38(28), 8849-8857 (1999-07-22)
Ligand-induced glucocorticoid receptor (GR) activation has recently been linked to the inhibition of cell proliferation via the transcriptional induction of p21(WAF1/Cip1), which functions as a universal inhibitor of cyclin-dependent protein kinases. Herein, we identify a Ser/Thr protein phosphatase (PP5) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service