Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9669

Sigma-Aldrich

Monoclonal Anti-Phycoerythrin antibody produced in mouse

clone PE-85, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PE-85, monoclonal

contains

15 mM sodium azide

species reactivity

(Porphyra tenera), (Porphyridium cruentum)

technique(s)

indirect ELISA: 1:2,000
western blot: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Specificity

The antibody reacts with R-phycoerythrin from Porphyra tenera and with B-phycoerythrin from Porphyridium cruentum. The antibody may be used in competitive ELISA and amplification of immunofluorescent labeling of mouse monoclonal antibodies with phycoerythrin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service