P9664
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt
solution, 10 mg/mL in chloroform, ≥99%
Synonym(s):
(9Z)-9-Octadecenoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester monosodium salt, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dioleoyl, L-α-Phosphatidyl-DL-glycerol, dioleoyl sodium salt, DOPG-Na, PG(18:1(9Z)/18:1(9Z))
About This Item
Recommended Products
Quality Level
assay
≥99%
form
solution
concentration
10 mg/mL in chloroform
lipid type
phosphoglycerides
storage temp.
−20°C
SMILES string
[Na].CCCCCCCC\C=C\CCCCCCCC(=O)OCC(COP(O)(=O)OCC(O)CO)OC(=O)CCCCCCC\C=C\CCCCCCCC
InChI
1S/C42H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h17-20,39-40,43-44H,3-16,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b19-17-,20-18-;/t39?,40-;/m1./s1
InChI key
IUVFCFQZFCOKRC-IPKKNMRRSA-M
Looking for similar products? Visit Product Comparison Guide
Other Notes
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service