Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9618

Sigma-Aldrich

pT7-FLAG-3 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shipped in

dry ice

storage temp.

−20°C

Application

Cytoplasmic expression of Met-N-terminal FLAG® fusion proteins under the T7/lacO promoter. The presence of both the kanamycin resistance gene and the ampicillin resistance gene offers higher flexibility of selection. Supplied with a pT7-FLAG-3-BAP Control Vector.

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service