Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9539

Sigma-Aldrich

Phytic acid calcium salt

≥90%

Synonym(s):

myo-Inositol hexakis(dihydrogen phosphate), Inositol hexakisphosphate, Inositol hexaphosphoric acid, InsP6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16CaO24P6
CAS Number:
Molecular Weight:
698.10
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥90%

form

powder

storage condition

desiccated

SMILES string

[Mg++].[Ca++].OP(O)(=O)O[C@@H]1[C@@H](OP(O)(O)=O)[C@@H](OP(O)([O-])=O)[C@@H](OP(O)([O-])=O)[C@H](OP(O)([O-])=O)[C@H]1OP(O)([O-])=O

Biochem/physiol Actions

Phytic acid is present in all eukaryotic cells. In plants, it is known to function as a [PO4]3− storage depot and precursor for other inositol phosphates and pyrophosphates. Its high surface negative charge makes it a potent chelator of divalent and trivalent cations in vitro, and it is most likely associated with Ca2+ or Mg2+ ions in vivo. In yeast, phytic acid acts with the nuclear pore protein Gle1 to regulate the nuclear export of mRNA by coactivating the RNA-dependent ATPase activity of DExD-box protein 5 (Dbp5).

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service