P9529
Phosphodiesterase, 3′,5′-cyclic-nucleotide-specific from bovine brain
lyophilized powder
Synonym(s):
3′,5′-Cyclic-nucleotide 5′-nucleotidohydrolase from bovine brain, PDE
About This Item
Recommended Products
biological source
bovine brain
form
lyophilized powder
specific activity
1-5 units/mg protein (without added calmodulin)
mol wt
~60 kDa
composition
Protein, 4.0-6.0% Lowry
UniProt accession no.
foreign activity
2′,3′-cyclic nucleotide phosphodiesterase, 5′-nucleotidase and alkaline phosphatase ≤0.1%
5′-ATPase ≤0.2%
inorganic pyrophosphatase ≤0.8%
storage temp.
−20°C
Gene Information
cow ... PDE1A(281969)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Packaging
Unit Definition
Physical form
Preparation Note
Reconstitution
Analysis Note
inhibitor
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service