Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9306

Sigma-Aldrich

Poly(Arg, Pro, Thr) hydrochloride

Arg:Pro:Thr (1:1:1), mol wt 5,000-20,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

form

solid

feed ratio

Arg:Pro:Thr (1:1:1)

mol wt

5,000-20,000

impurities

<5% Ornithine residues

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Polymer composition may deviate from the molar feed ratios. Molecular weight based on viscosity of precursor ornithine copolymer.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Serizawa et al.
Langmuir : the ACS journal of surfaces and colloids, 25(20), 12229-12234 (2009-09-23)
A novel synthetic route to peptide-capped gold nanoparticles was demonstrated herein. Tetrachloroaurate ions were reduced with 2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulfonic acid (HEPES) under extremely mild conditions (pH 7.2, ambient temperature) in the presence of cysteine-terminal desired peptides, so that peptide-capped spherical nanoparticles were

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service