Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P9305

Sigma-Aldrich

Phosphate-Citrate Buffer with Urea Hydrogen Peroxide

tablet

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161700

form

tablet

reaction suitability

reagent type: oxidant

solubility

water: 1 tablet/100mL, clear, colorless

Storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Phosphate-Citrate Buffer Tablets with Urea Hydrogen Peroxide may be used as a peroxidase substrate buffer for ELISA with the H2O2 included.

Reconstitution

One tablet dissolved in 100 mL of deionized water yields a 0.05 M phosphate-citrate buffer, containing 0.014% H2O2, pH 5.0 at 25 °C.

Pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John-Paul Westwood et al.
British journal of haematology, 164(6), 858-866 (2014-01-01)
Complement dysregulation is key in the pathogenesis of atypical Haemolytic Uraemic Syndrome (aHUS), but no clear role for complement has been identified in Thrombotic Thrombocytopenic Purpura (TTP). We aimed to assess complement activation and cytokine response in acute antibody-mediated TTP.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service